1,2-Diphenylethane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:103-29-7
:
Formula:(C₆H₅)CH₂CH₂(C₆H₅)
MDL Number:MFCD00004796
:182.27 g/mol
Merck Index:14,01198
Boiling Pt:284 °C (1013 hPa)
Melting Pt:52…54 °C
Density:1,12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-096-4
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:50.0 to 53.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together