

4,4'-Bipyridyl ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:553-26-4
:
Formula:C₁₀H₈N₂
MDL Number:MFCD00006416
:156.19 g/mol
Merck Index:14,03348
Boiling Pt:161°C
Melting Pt:78 °C
Storage Temperature:Ambient
EINECS:209-036-3
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:111.0 to 114.0 deg-C
Specification Test Results
Frequently Bought Together