

1-Methylimidazole (NMI) ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:616-47-7
:
Formula:C₄H₆N₂
MDL Number:MFCD00005292
Boiling Pt:198°C
Melting Pt:–2 °C
Density:1,03 g/cm³ (20 °C)
Flash Pt:92°C
Storage Temperature:Ambient
EINECS:210-484-7
ADR:8,III
:82.11 g/mol
UN:1760
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Related Information
Frequently Bought Together