
4,4'-Dipyridyl disulphide ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2645-22-9
:
Formula:C₁₀H₈N₂S₂
MDL Number:MFCD00006423
:220.32 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:155 °C
Storage Temperature:Refrigerator
EINECS:220-158-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:74.0 to 79.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together