



Resorcinol ≥99.0% (by GC)
: TCI
:108-46-3
:
:C₆H₆O₂
:MFCD00002269
:14,08155
:281 °C (1013 hPa)
:109…111 °C
:1,272 g/cm³ (20 °C)
: (closed cup)
:2876
:6.1,III
:110.11 g/mol
:203-585-2
:GHS05
:GHS06
:GHS08
:GHS09
:Ambient
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:110.0 to 112.0 deg-C
- Solubility in Water:almost transparency
Frequently Bought Together