


Potassium-tert-butoxide ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:865-47-4
:
Formula:C₄H₉KO
MDL Number:MFCD00012162
:112.21 g/mol
Melting Pt:256…258 °C
EINECS:212-740-3
UN:3206
ADR:4.2,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together