
3-Methyl-2-pyridone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1003-56-1
:91914-04-4
Formula:C₆H₇NO
MDL Number:MFCD00039704
:109.13 g/mol
EINECS:213-710-2
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:140.0 to 144.0 deg-C
Specification Test Results
Frequently Bought Together