Tetraethyl orthosilicate (TEOS) ≥97.0% (by GC)
: TCI
:78-10-4
:
:Si(OC₂H₅)₄
:MFCD00009062
:208.33 g/mol
:14,03851
:165°C
:–85.5 °C
:0,933 g/cm³ (20 °C)
:54°C
:Ambient
:201-083-8
:1292
:3,III
:GHS02
:GHS07
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together