Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
2-Ethoxyethanol ≥99.0% (by GC)
 :  TCI
 
undefined
 
 
 
2-Ethoxyethanol ≥99.0% (by GC)
 :  TCI
 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

Frequently Bought Together