(±)-1-Aminopropan-2-ol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:78-96-6
:
Formula:C₃H₉NO
MDL Number:MFCD00008139
:75.11 g/mol
Boiling Pt:160°C
Melting Pt:–2°C
Density:0,962 g/cm³ (20 °C)
Flash Pt:78°C
Storage Temperature:Ambient
EINECS:201-162-7
UN:2735
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together