
2-(Trifluoromethoxy)benzaldehyde ≥95.0% (by GC)
TCIAT2302-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
:
:190.12 g/mol
:76°C
:67°C
:GHS07
:94651-33-9
:C₈H₅F₃O₂
:MFCD00042405