
2-Naphthylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:581-96-4
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00004126
:186.21 g/mol
Boiling Pt:352 °C (1013 hPa)
Melting Pt:139…142 °C
Density:1,29 g/cm³ (20 °C)
EINECS:209-475-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:142.0 to 145.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together