


Acetyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:75-36-5
:
Formula:C₂H₃ClO
MDL Number:MFCD00000719
:78.5 g/mol
Merck Index:14,00085
Boiling Pt:52°C
Melting Pt:–112°C
Density:1,1 g/cm³ (20 °C)
Flash Pt:5°C
Storage Temperature:Refrigerator
EINECS:200-865-6
UN:1717
ADR:3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together