
Sodium dehydracetate monohydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0040-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:8.3 to 10.0 %
Specification Test Results
About this item
:
:208.15 g/mol
EINECS:224-580-1
:GHS07
CAS Number:64039-28-7
:MFCD01694098
Formula:C₈H₇NaO₄·1H₂O
MDL Number:MFCD00040583