
2,4-Dichlorobenzylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95-00-1
:
Formula:C₇H₇Cl₂N
MDL Number:MFCD00008109
:176.04 g/mol
Boiling Pt:83 °C (1013 hPa)
Density:1,33 g/cm³ (20 °C)
EINECS:202-382-6
UN:1760
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together