

3,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:102-36-3
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002017
Boiling Pt:234 °C (1013 hPa)
Melting Pt:38…41 °C
Storage Temperature:Refrigerator
EINECS:203-026-2
UN:2250
ADR:6.1,II
:188.01 g/mol
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Butylamine Method):min. 98.0 %
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together