Order Entry
Italy
ContactUsLinkComponent
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥99.0% (by GC)
Corrosive
Irritant
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥99.0% (by GC)
Catalog # TCIAC1036-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥99.0% (by GC)
Catalog # TCIAC1036-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (GC):
    min. 99.0 %
  • Melting point:
    62.0 to 64.0 deg-C

Specification Test Results

About this item

 :
 :270.55 g/mol
Melting Pt:60…65 °C
EINECS:206-883-0
ADR:8,III
 :GHS05
 :GHS07
CAS Number:392-95-0
Formula:C₇H₂ClF₃N₂O₄
MDL Number:MFCD00007076