
1,5-Dichloro-9,10-anthraquinone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:82-46-2
:
Formula:C₁₄H₆Cl₂O₂
MDL Number:MFCD00001190
:277.11 g/mol
Boiling Pt:401 °C (1013 hPa)
Melting Pt:245…250 °C
Density:1,63 g/cm³ (20 °C)
EINECS:201-424-0
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:248.0 to 253.0 deg-C
- Solubility in hot Nitorobenzen:very faint turbidity
Specification Test Results
Frequently Bought Together