
1,2-Di(2-pyridyl)ethylene ≥98.0% (by GC, titration analysis)
TCIAB1131-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:119.0 to 122.0 deg-C
- Solubility in Methanol:almost transparency
:
:347 °C (1013 hPa)
:119 °C
:182.22 g/mol
:215-866-7
:GHS07
:1437-15-6
:C₁₂H₁₀N₂
:MFCD00006356