

1,2-Dimethylimidazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1739-84-0
:
Formula:C₅H₈N₂
MDL Number:MFCD00005294
:96.13 g/mol
Boiling Pt:205°C
Melting Pt:38 °C
Density:1,084 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-101-2
UN:3263
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together