1,3-Dithiane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:505-23-7
:
Formula:C₄H₈S₂
MDL Number:MFCD00006654
:120.24 g/mol
Boiling Pt:208°C
Melting Pt:51…54 °C
Density:1,36 g/cm³ (25 °C)
EINECS:208-006-7
ADR:9,
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:52.0 to 55.0 deg-C
Specification Test Results
Frequently Bought Together