
2,3-Dimethylquinoxaline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2379-55-7
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00006728
:158.2 g/mol
Boiling Pt:130°C
Density:1,226 g/cm³ (20 °C)
EINECS:219-162-0
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:104.0 to 109.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together