Order Entry
Italy
ContactUsLinkComponent
7-Isopropyl-1,4-dimethylazulene ≥99.0% (by GC)
 
7-Isopropyl-1,4-dimethylazulene ≥99.0% (by GC)
  TCIAG0228-10G
 :  TCI
 :  
undefined
 
7-Isopropyl-1,4-dimethylazulene ≥99.0% (by GC)
  TCIAG0228-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Dark blue powder to lump
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :198.31 g/mol
 :14,04554
 :320°C
 :133 °C
 :0,9759 g/cm³ (18 °C)
 :207-701-2
 :GHS07
 :489-84-9
 :C₁₅H₁₈
 :MFCD00003811