

1,3-Dioxane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:505-22-6
:
Formula:C₄H₈O₂
MDL Number:MFCD00006566
:88.11 g/mol
Boiling Pt:105°C
Melting Pt:–74.2 °C
Density:1,0399 g/cm³ (15 °C)
Flash Pt:1°C
Storage Temperature:Ambient
EINECS:208-005-1
UN:1165
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together