Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethylanthracene ≥98.0% (by GC)
2,3-Dimethylanthracene ≥98.0% (by GC)
Catalog # TCIAD2979-200MG
Supplier:  TCI
CAS Number:  
undefined
2,3-Dimethylanthracene ≥98.0% (by GC)
Catalog # TCIAD2979-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :206.29 g/mol
Melting Pt:252°C
EINECS:210-328-8
CAS Number:613-06-9
Formula:C₁₆H₁₄
MDL Number:MFCD00154982