


2-Butyne-1,4-diol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:110-65-6
:
Formula:C₄H₆O₂
MDL Number:MFCD00002915
:86.09 g/mol
Boiling Pt:238°C
Melting Pt:55…58 °C
Density:1,1001 g/cm³ (20 °C)
Flash Pt:142 °C
Storage Temperature:Ambient
EINECS:203-788-6
UN:2716
ADR:6.1,III
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:54.0 to 58.0 deg-C
- Solubility in Methanol:within almost transparency
Specification Test Results
Frequently Bought Together