2,5-Dimethyl-2,5-hexanediol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:110-03-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004473
:146.23 g/mol
Boiling Pt:127°C
Melting Pt:85…88 °C
Density:0,898 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-731-5
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:87.0 to 92.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together