
(R)-(-)-1,2-Propanediol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4254-14-2
:
Formula:C₃H₈O₂
MDL Number:MFCD00066248
:76.1 g/mol
Merck Index:14,07855
Flash Pt:103°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
- Optical purity (GC):min. 97.0 ee%
- Specific rotation [a]20/D:-15.0 to -18.0 deg (neat)
Specification Test Results
Frequently Bought Together