Order Entry
Italy
ContactUsLinkComponent
 
2-Mercaptoethanol ≥98.0% (by GC)
 :  TCI
 
undefined
2-Mercaptoethanol ≥98.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together