Order Entry
Italy
ContactUsLinkComponent
2-(Methylthio)phenol ≥97.0% (by GC, titration analysis)
Irritant
2-(Methylthio)phenol ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1854-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-(Methylthio)phenol ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1854-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :140.21 g/mol
Boiling Pt:105°C
Density:1,168 g/cm³ (25 °C)
EINECS:214-027-2
 :GHS07
CAS Number:1073-29-6
Formula:C₇H₈OS
MDL Number:MFCD00002211