
1-Methyl-2-pyridone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:694-85-9
:
Formula:C₆H₇NO
MDL Number:MFCD00006520
:109.13 g/mol
Boiling Pt:252°C
Melting Pt:33 °C
Density:1,1321 g/cm³ (18 °C)
EINECS:211-777-2
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together