Benzoic anhydride ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:93-97-0
:
Formula:C₁₄H₁₀O₃
MDL Number:MFCD00003073
Merck Index:14,01092
Boiling Pt:158°C
Melting Pt:40 °C
Density:1,231 to 1.247 g/cm³ (4 °C)
:226.23 g/mol
EINECS:202-291-1
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Morpholine Method):min. 97.0 %
- Melting point:41.0 to 44.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together