Order Entry
Italy
ContactUsLinkComponent
 
 
 
(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -20.0 to -24.0 deg (C=10, CHCl3)

 

Frequently Bought Together