

Dimethyl sulphoxide ≥99.0% (by GC)
: TCI
:67-68-5
:
:(CH₃)₂SO
:MFCD00002089
:78.14 g/mol
:14,03259
:189°C
:18.5 °C
:1,101 g/cm³ (20 °C)
:95°C
:Ambient
:200-664-3
:GHS07
:GHS08
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together