

2,3-Diaminotoluene ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2687-25-4
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00011589
:122.17 g/mol
Boiling Pt:225°C
Melting Pt:113 °C
Density:1,3 g/cm³ (20 °C)
EINECS:220-248-5
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:58.0 to 62.0 deg-C
Specification Test Results
Frequently Bought Together