

1-Naphthyl isothiocyanate ≥98.0% (by GC)
: TCI
:551-06-4
:
:C₁₁H₇NS
:MFCD00003882
:185.25 g/mol
:14,06411
:100 °C (0,2 -rr)
:59 °C
:1,81 g/cm³ (20 °C)
:208-990-8
:2811
:6.1,III
:GHS06
:GHS08
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:53.0 to 57.0 deg-C
- Solubility in hot EtOH:very faint turbidity
Frequently Bought Together