
2-Iodobenzoyl chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:609-67-6
:
Formula:C₇H₄ClIO
MDL Number:MFCD00001040
:266.47 g/mol
Boiling Pt:106°C
Melting Pt:27…30 °C
Storage Temperature:Ambient
EINECS:210-196-1
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to lump
- Purity (GC):min. 95.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together