Order Entry
Italy
ContactUsLinkComponent
 
Irritant
3-Amino-4-fluorobenzoic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAA1972-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-Amino-4-fluorobenzoic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAA1972-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    182.0 to 186.0 deg-C

Specification Test Results

About this item

 :
 :155.13 g/mol
EINECS:219-122-2
 :GHS07
CAS Number:2365-85-7
Formula:C₇H₆FNO₂
MDL Number:MFCD00130055