Order Entry
Italy
ContactUsLinkComponent
α,α,α-Trifluoro-o-toluic acid ≥98.0% (by HPLC, titration analysis)
Irritant
α,α,α-Trifluoro-o-toluic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1302-10G
Supplier:  TCI
CAS Number:  
undefined
Irritant
α,α,α-Trifluoro-o-toluic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1302-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    107.0 to 111.0 deg-C

Specification Test Results

About this item

 :
Boiling Pt:247°C
Melting Pt:110 °C
 :190.12 g/mol
EINECS:207-093-9
 :GHS07
CAS Number:433-97-6
Formula:C₈H₅F₃O₂
MDL Number:MFCD00002476