Maleic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-16-7
:
Formula:C₄H₄O₄
MDL Number:MFCD00063177
:116.07 g/mol
Merck Index:14,05703
Boiling Pt:256 °C (1013 hPa)
Melting Pt:131°C
Density:1,59 g/cm³ (20 °C)
Flash Pt: (closed cup)
Storage Temperature:Ambient
EINECS:203-742-5
UN:3261
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Water:transparency
Specification Test Results
Frequently Bought Together