
Phthalic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:88-99-3
:
Formula:C₈H₆O₄
MDL Number:MFCD00002467
:166.13 g/mol
Merck Index:14,07371
Boiling Pt:315 - 317 °C (1013 hPa)
Melting Pt:210°C
Density:1,576 g/cm³ (20 °C)
EINECS:201-873-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together