
2,2-Dimethylglutaric acid ≥98.0% (by GC, titration analysis)
TCIAD1786-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:83.0 to 87.0 deg-C
- Solubility in Water:very faint turbidity
:
:160.17 g/mol
:200°C
:83…84 °C
:211-655-9
:GHS07
:Ambient
:681-57-2
:C₇H₁₂O₄
:MFCD00004197