Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Mercaptopropionic acid ≥98.0% (by GC, titration analysis)

3-Mercaptopropionic acid ≥98.0% (by GC, titration analysis)

Corrosive
Toxic
Supplier:   TCI
3-Mercaptopropionic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
3-Mercaptopropionic acid ≥98.0% (by GC, titration analysis)
3-Mercaptopropionic acid ≥98.0% (by GC, titration analysis)

3 Options Available Below

Product Details & Documents
  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:116°C
  • Melting Pt:17…19 °C
  • Density:1,2199 g/cm³ (20 °C)
  • Flash Pt:132°C
  • Storage Temperature:Ambient
  • EINECS:203-537-0
  • UN:2922
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Iodometric Titration):
    min. 98.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 16,00 €
    16,00 €
    1 * 25 g
       16,00 €

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 20,00 €
    20,00 €
    1 * 100 g
       20,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 63,00 €
    63,00 €
    1 * 500 g
       63,00 €