Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Hydroxyurea ≥97.0% (by total nitrogen basis)

Health
Supplier:   TCI
Hydroxyurea ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Hydroxyurea ≥97.0% (by total nitrogen basis)

2 Options Available Below

Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:14,04848
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
  • UN:2811
  • ADR:6.1,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 18,00 €
    18,00 €
    1 * 5 g
       18,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 54,00 €
    54,00 €
    1 * 25 g
       54,00 €