Order Entry
Italy
ContactUsLinkComponent
1,5-Diamino-9,10-anthraquinone ≥92.0% (by total nitrogen basis)
Irritant
1,5-Diamino-9,10-anthraquinone ≥92.0% (by total nitrogen basis)
Catalog # TCIAD1268-5G
Supplier:  TCI
CAS Number:  
Irritant
1,5-Diamino-9,10-anthraquinone ≥92.0% (by total nitrogen basis)
Catalog # TCIAD1268-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystaline
  • Purity (HPLC):
    min. 85.0 area%
  • Purity (with Total Nitrogen):
    min. 92.0 %

Specification Test Results

About this item

 :
Melting Pt:314 °C
 :238.25 g/mol
EINECS:204-947-2
 :GHS07
CAS Number:129-44-2
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001226