Order Entry
Italy
ContactUsLinkComponent
 
2-Thiobarbituric acid ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
2-Thiobarbituric acid ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in Sodium hydroxide solution:
    almost transparency

 

Frequently Bought Together