

3-Butyne-2-one ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1423-60-5
:
Formula:C₄H₄O
MDL Number:MFCD00008775
:68.08 g/mol
Boiling Pt:86°C
Density:0,886 g/cm³ (15 °C)
Flash Pt:–1°C
Storage Temperature:Refrigerator
EINECS:215-834-2
UN:1992
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together