Order Entry
Italy
ContactUsLinkComponent
3,6-Dihydroxyphthalonitrile ≥97.0% (by GC, titration analysis)
Toxic
3,6-Dihydroxyphthalonitrile ≥97.0% (by GC, titration analysis)
Catalog # TCIAD1610-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
3,6-Dihydroxyphthalonitrile ≥97.0% (by GC, titration analysis)
Catalog # TCIAD1610-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Brown powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

About this item

 :
 :160.13 g/mol
Storage Temperature:Ambient
EINECS:225-241-0
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:4733-50-0
Formula:C₈H₄N₂O₂
MDL Number:MFCD00001790