


(±)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
: TCI
:598-78-7
:
:C₃H₅ClO₂
:MFCD00004224
:108.52 g/mol
:186°C
:–12°C
:1,27 g/cm³ (20 °C)
:106°C
:209-952-3
:2511
:8,III
:GHS05
:GHS06
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together