
4,7-Dichloroisatin ≥97.0% (by titrimetric analysis)
: TCI
:18711-13-2
:
:C₈H₃Cl₂NO₂
:MFCD00047214
:216.02 g/mol
:252°C
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 97.0 %
Frequently Bought Together